1-(2,6-Dichloro-3-(methylthio)phenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

273821-5G 273821-1G 273821-25G 273821-500MG
77628-346EA 1690.23 USD
77628-346 77637-664 77654-620 77645-632
1-(2,6-Dichloro-3-(methylthio)phenyl)-2,2,2-trifluoroethanone ≥95%
1-(2,6-Dichloro-3-(methylthio)phenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 289.102 g/mol
MW: C₉H₅Cl₂F₃OS
Storage Temperature: Ambient
MDL Number: MFCD34880682
CAS Number: 2969378-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR