(3-Bromo-2,4-dichlorophenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

273820-5G 273820-1G 273820-25G 273820-500MG
77632-390EA 1441 USD
77632-390 77637-662 77658-376 77645-630
(3-Bromo-2,4-dichlorophenyl)(methyl)sulfane ≥95%
(3-Bromo-2,4-dichlorophenyl)(methyl)sulfane

Precaución: For Research Use Only

Formula: C₇H₅BrCl₂S
Storage Temperature: Ambient
MDL Number: MFCD34880681
CAS Number: 1672665-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR