1-Bromo-2-fluoro-3-(piperidinomethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

27364-5G 27364-1G 27364-500MG 27364-25G
77634-580EA 342 USD
77634-580 77637-660 77652-558 77658-374
1-Bromo-2-fluoro-3-(piperidinomethyl)benzene ≥95%
1-Bromo-2-fluoro-3-(piperidinomethyl)benzene

Precaución: For Research Use Only

Formula: 272.157 g/mol
MW: C₁₂H₁₅BrFN
Storage Temperature: Ambient
MDL Number: MFCD21333072
CAS Number: 1355246-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR