Ethyl 6-(pyrrolidin-1-yl)nicotinate ≥95%

Supplier: AOBChem USA

Warning

27359-5G 27359-1G 27359-25G 27359-500MG
77632-388EA 628 USD
77632-388 77637-658 77654-618 77645-628
Ethyl 6-(pyrrolidin-1-yl)nicotinate ≥95%
Ethyl 6-(pyrrolidin-1-yl)nicotinate

Precaución: For Research Use Only

Formula: 220.268 g/mol
MW: C₁₂H₁₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11616057
CAS Number: 897399-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR