(4-Chloro-2-(cyclopropylmethoxy)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

273195-5G 273195-1G 273195-25G 273195-500MG
77632-386EA 1441 USD
77632-386 77637-656 77654-614 77645-626
(4-Chloro-2-(cyclopropylmethoxy)phenyl)(methyl)sulfane ≥95%
(4-Chloro-2-(cyclopropylmethoxy)phenyl)(methyl)sulfane

Precaución: For Research Use Only

Formula: 228.738 g/mol
MW: C₁₁H₁₃ClOS
Storage Temperature: Ambient
MDL Number: MFCD34880185
CAS Number: 2969379-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR