3-(3-Chloro-2-fluorophenyl)tetrahydrofuran-3-ol ≥97%

Supplier: AOBChem USA

Warning

272418-5G 272418-1G 272418-25G 272418-500MG
77628-344EA 1484.23 USD
77628-344 77637-652 77658-370 77645-624
3-(3-Chloro-2-fluorophenyl)tetrahydrofuran-3-ol ≥97%
3-(3-Chloro-2-fluorophenyl)tetrahydrofuran-3-ol

Precaución: For Research Use Only

Formula: 216.6366 g/mol
MW: C₁₀H₁₀ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD34879666
CAS Number: 2969460-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR