3-Bromo-N,N,4-trimethylaniline ≥97%

Supplier: AOBChem USA

Warning

27228-5G 27228-1G 27228-25G 27228-500MG
77634-578EA 573 USD
77634-578 77637-650 77654-612 77649-708
3-Bromo-N,N,4-trimethylaniline ≥97%
3-Bromo-N,N,4-trimethylaniline

Precaución: For Research Use Only

Formula: 214.102 g/mol
MW: C₉H₁₂BrN
Storage Temperature: Ambient
MDL Number: MFCD14529603
CAS Number: 53104-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR