1,2,3,5-Tetrabromo-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

272060-5G 272060-1G 272060-25G 272060-500MG
77628-342EA 1484.23 USD
77628-342 77637-648 77654-610 77649-706
1,2,3,5-Tetrabromo-4-methylbenzene ≥97%
1,2,3,5-Tetrabromo-4-methylbenzene

Precaución: For Research Use Only

Formula: 407.72 g/mol
MW: C₇H₄Br₄
Storage Temperature: Ambient
MDL Number: MFCD35168869
CAS Number: 39664-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR