4-Fluoro-2-n-propoxybenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
27168-5G
27168-1G
27168-25G
27168-500MG
77634-576EA
564
USD
77634-576
77637-646
77654-608
77649-704
4-Fluoro-2-n-propoxybenzoic acid ≥97%
4-Fluoro-2-n-propoxybenzoic acid
Precaución: For Research Use Only
Formula:
C₁₀H₁₁FO₃ Storage Temperature: Ambient |
MDL Number:
MFCD14582792 CAS Number: 1378666-16-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...