3-Bromo-2-methoxy-6-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

270905-1G 270905-5G 270905-500MG
77637-640EA 649 USD
77637-640 77658-362 77645-618
3-Bromo-2-methoxy-6-methylbenzaldehyde ≥95%
3-Bromo-2-methoxy-6-methylbenzaldehyde

Precaución: For Research Use Only

Formula: 229.07 g/mol
MW: C₉H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD28348646
CAS Number: 1782219-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR