3-Bromo-2-methoxy-6-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

270904-1G 270904-5G 270904-500MG
77634-570EA 1156 USD
77634-570 77654-606 77645-616
3-Bromo-2-methoxy-6-methylbenzoic acid ≥97%
3-Bromo-2-methoxy-6-methylbenzoic acid

Precaución: For Research Use Only

Formula: C₉H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD01846085
CAS Number: 681467-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR