3-Bromo-5-fluoro-N,N-diethylaniline ≥97%

Supplier: AOBChem USA

Warning

27081-5G 27081-25G 27081-1G 27081-500MG
77632-380EA 841 USD
77632-380 77658-360 77641-654 77649-698
3-Bromo-5-fluoro-N,N-diethylaniline ≥97%
3-Bromo-5-fluoro-N,N-diethylaniline

Precaución: For Research Use Only

Formula: C₁₀H₁₃BrFN
Storage Temperature: Ambient
MDL Number: MFCD28142588
CAS Number: 1809161-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR