Ethyl 5-bromo-4-methylpicolinate ≥97%

Supplier: AOBChem USA

Warning

27055-5G 27055-25G 27055-1G 27055-500MG
77632-378EA 656 USD
77632-378 77654-604 77641-652 77645-614
Ethyl 5-bromo-4-methylpicolinate ≥97%
Ethyl 5-bromo-4-methylpicolinate

Precaución: For Research Use Only

Formula: 244.085 g/mol
MW: C₉H₁₀BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD21604170
CAS Number: 1122090-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR