Methyl 6-bromo-4-methylpyridine-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

27046-5G 27046-25G 27046-1G 27046-500MG
77632-376EA 1209 USD
77632-376 77654-602 77641-650 77649-696
Methyl 6-bromo-4-methylpyridine-3-carboxylate ≥97%
Methyl 6-bromo-4-methylpyridine-3-carboxylate

Precaución: For Research Use Only

Formula: 230.059 g/mol
MW: C₈H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD21087967
CAS Number: 1355229-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR