1,4-Bis(methylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

270435-5G 270435-25G 270435-1G 270435-500MG
77632-374EA 1268 USD
77632-374 77658-358 77641-648 77645-612
1,4-Bis(methylthio)benzene ≥95%
1,4-Bis(methylthio)benzene

Precaución: For Research Use Only

Formula: 170.3 g/mol
MW: C₈H₁₀S₂
Storage Temperature: Ambient
MDL Number: MFCD00465085
CAS Number: 699-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR