(5-Chloro-2-ethoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

27008-5G 27008-1G 27008-25G 27008-500MG
77634-568EA 609 USD
77634-568 77637-638 77654-600 77645-610
(5-Chloro-2-ethoxyphenyl)methanol ≥97%
(5-Chloro-2-ethoxyphenyl)methanol

Precaución: For Research Use Only

Formula: 186.635 g/mol
MW: C₉H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD11103200
CAS Number: 23426-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR