1,3-Dichloro-2,4,6-trifluorobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,4-Dichloro-1,3,5-trifluorobenzene

269391-5G 269391-25G
77634-566EA 119 USD
77634-566 77654-596
1,3-Dichloro-2,4,6-trifluorobenzene ≥97%
1,3-Dichloro-2,4,6-trifluorobenzene

Precaución: For Research Use Only

Formula: C₆HCl₂F₃
MW: 200.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012242
CAS Number: 2368-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR