2-Flouro-4-(methylthio)pyridine-3-boronic acid ≥95%

Supplier: AOBChem USA

Warning

26938-1G 26938-5G 26938-500MG
77637-634EA 588 USD
77637-634 77654-594 77649-694
2-Flouro-4-(methylthio)pyridine-3-boronic acid ≥95%
2-Flouro-4-(methylthio)pyridine-3-boronic acid

Precaución: For Research Use Only

Formula: 187 g/mol
MW: C₆H₇BFNO₂S
Storage Temperature: Ambient
MDL Number: MFCD27756689
CAS Number: 2121513-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR