1-Fluoro-4-[(4-iodophenyl)thio]benzene ≥95%

Supplier: AOBChem USA

Warning

269204-5G 269204-1G 269204-25G 269204-500MG
77628-334EA 1484.23 USD
77628-334 77637-632 77658-354 77645-604
1-Fluoro-4-[(4-iodophenyl)thio]benzene ≥95%
1-Fluoro-4-[(4-iodophenyl)thio]benzene

Precaución: For Research Use Only

Formula: 330.16 g/mol
MW: C₁₂H₈FIS
Storage Temperature: Ambient
MDL Number: MFCD22488507
CAS Number: 143128-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR