4-[(4-Fluorophenyl)sulfanyl]aniline ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-(4-Fluorophenylthio)aniline

269173-5G 269173-25G 269173-1G 269173-500MG
77634-562EA 449 USD
77634-562 77654-590 77641-644 77649-692
4-[(4-Fluorophenyl)sulfanyl]aniline ≥95%
4-[(4-Fluorophenyl)sulfanyl]aniline

Precaución: For Research Use Only

Formula: C₁₂H₁₀FNS
MW: 219.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00172068
CAS Number: 24900-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR