3-Bromo-5-fluoro-N,N-dimethylaniline ≥97%

Supplier: AOBChem USA

Warning

26917-5G 26917-1G 26917-25G 26917-500MG
77634-564EA 400 USD
77634-564 77637-630 77658-352 77645-602
3-Bromo-5-fluoro-N,N-dimethylaniline ≥97%
3-Bromo-5-fluoro-N,N-dimethylaniline

Precaución: For Research Use Only

Formula: C₈H₉BrFN
Storage Temperature: Ambient
MDL Number: MFCD27756687
CAS Number: 1129542-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR