N-(4-chloro-2-formylphenyl)-2,2-dimethylpropionamide ≥97%
Supplier: AOBChem USA
|
Warning
|
26888-5G
26888-1G
26888-25G
26888-500MG
77634-560EA
295
USD
77634-560
77637-628
77654-586
77649-690
N-(4-chloro-2-formylphenyl)-2,2-dimethylpropionamide ≥97%
N-(4-chloro-2-formylphenyl)-2,2-dimethylpropionamide
Precaución: For Research Use Only
Formula:
C₁₂H₁₄ClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD27756683 CAS Number: 127472-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...