1-Bromo-3,5-bis(1-methylethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

268646-5G 268646-1G 268646-500MG
77628-332EA 1484.23 USD
77628-332 77637-624 77645-598
1-Bromo-3,5-bis(1-methylethoxy)benzene ≥97%
1-Bromo-3,5-bis(1-methylethoxy)benzene

Precaución: For Research Use Only

Formula: 273.17 g/mol
MW: C₁₂H₁₇BrO₂
Storage Temperature: Ambient
MDL Number: MFCD28356585
CAS Number: 1310418-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR