2-Fluoro-5-phenylpyridine-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

26858-5G 26858-25G 26858-1G 26858-500MG
77632-370EA 1198 USD
77632-370 77654-584 77641-640 77645-596
2-Fluoro-5-phenylpyridine-3-carboxylic acid ≥97%
2-Fluoro-5-phenylpyridine-3-carboxylic acid

Precaución: For Research Use Only

Formula: C₁₂H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD14701978
CAS Number: 1214349-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR