3-(2,5-Difluorophenyl)tetrahydro-3-furanol ≥95%

Supplier: AOBChem USA

Warning

268572-5G 268572-1G 268572-500MG
77628-330EA 1484.23 USD
77628-330 77637-622 77645-594
3-(2,5-Difluorophenyl)tetrahydro-3-furanol ≥95%
3-(2,5-Difluorophenyl)tetrahydro-3-furanol

Precaución: For Research Use Only

Formula: 200.18 g/mol
MW: C₁₀H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18332670
CAS Number: 1342055-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR