3-(2,4-Difluorophenyl)tetrahydro-3-furanol ≥95%

Supplier: AOBChem USA

Warning

268569-5G 268569-1G 268569-500MG
77628-328EA 1484.23 USD
77628-328 77637-620 77645-592
3-(2,4-Difluorophenyl)tetrahydro-3-furanol ≥95%
3-(2,4-Difluorophenyl)tetrahydro-3-furanol

Precaución: For Research Use Only

Formula: 200.18 g/mol
MW: C₁₀H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18332669
CAS Number: 1343023-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR