3-(2,4-Dichlorophenyl)tetrahydro-3-furanol ≥95%

Supplier: AOBChem USA

Warning

268560-500MG 268560-5G 268560-1G
77652-554EA 379 USD
77652-554 77654-582 77641-638
3-(2,4-Dichlorophenyl)tetrahydro-3-furanol ≥95%
3-(2,4-Dichlorophenyl)tetrahydro-3-furanol

Precaución: For Research Use Only

Formula: C₁₀H₁₀Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34878112
CAS Number: 1505346-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR