3-(5-Bromo-2-fluorophenyl)tetrahydro-3-furanol ≥95%

Supplier: AOBChem USA

Warning

268557-5G 268557-1G 268557-500MG
77654-580EA 2073 USD
77654-580 77641-636 77649-684
3-(5-Bromo-2-fluorophenyl)tetrahydro-3-furanol ≥95%
3-(5-Bromo-2-fluorophenyl)tetrahydro-3-furanol

Precaución: For Research Use Only

Formula: 261.09 g/mol
MW: C₁₀H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD23759103
CAS Number: 1519588-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR