4-Chloro-2-iodo-pivaloylaniline ≥97%

Supplier: AOBChem USA

Warning

26844-5G 26844-1G 26844-25G 26844-500MG
77632-368EA 636 USD
77632-368 77637-612 77658-346 77645-584
4-Chloro-2-iodo-pivaloylaniline ≥97%
4-Chloro-2-iodo-pivaloylaniline

Precaución: For Research Use Only

Formula: 337.584 g/mol
MW: C₁₁H₁₃ClINO
Storage Temperature: Ambient
MDL Number: MFCD27756678
CAS Number: 1809161-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR