Tetrahydro-3-(3-methylphenyl)-3-furanol ≥95%

Supplier: AOBChem USA

Warning

268389-5G 268389-25G 268389-1G 268389-500MG
77628-316EA 1484.23 USD
77628-316 77635-572 77637-608 77645-582
Tetrahydro-3-(3-methylphenyl)-3-furanol ≥95%
Tetrahydro-3-(3-methylphenyl)-3-furanol

Precaución: For Research Use Only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD18332713
CAS Number: 1343279-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR