4-(Difluoromethyl)benzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-(Difluoromethyl)phenyl)boronic acid, 4-Boronobenzal fluoride

26801-5G 26801-25G
77634-558EA 88 USD
77634-558 77654-576
4-(Difluoromethyl)benzeneboronic acid ≥97%
4-(Difluoromethyl)benzeneboronic acid

Precaución: For Research Use Only

Formula: C₇H₇BF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09056705
CAS Number: 946525-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR