1-(5-Methyl-3-thienyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

267913-1G 267913-5G 267913-500MG
77637-606EA 593 USD
77637-606 77654-574 77645-580
1-(5-Methyl-3-thienyl)ethanone ≥95%
1-(5-Methyl-3-thienyl)ethanone

Precaución: For Research Use Only

Formula: 140.2 g/mol
MW: C₇H₈OS
Storage Temperature: Ambient
MDL Number: MFCD21120473
CAS Number: 69213-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR