4-(Pyridin-4-yl)phenyl boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-(Pyridin-4-yl)phenyl)boronic acid

26791-5G 26791-25G
77634-556EA 69 USD
77634-556 77654-572
4-(Pyridin-4-yl)phenyl boronic acid ≥97%
4-(Pyridin-4-yl)phenyl boronic acid

Precaución: For Research Use Only

Formula: C₁₁H₁₀BNO₂
MW: 199.014 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17170043
CAS Number: 1045332-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR