2-Bromo-4-chlorothiophene ≥97%

Supplier: AOBChem USA

Warning

26770-1G 26770-5G 26770-500MG
77637-604EA 775 USD
77637-604 77658-344 77645-578
2-Bromo-4-chlorothiophene ≥97%
2-Bromo-4-chlorothiophene

Precaución: For Research Use Only

Formula: 197.481 g/mol
MW: C₄H₂BrClS
Storage Temperature: Ambient
MDL Number: MFCD26520773
CAS Number: 32431-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR