Ethyl 1,3,5-trimethyl-1H-pyrazole-4-carboxylate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(Ethoxycarbonyl)-1,3,5-trimethyl-1H-pyrazole

267384-5G 267384-1G 267384-25G
77634-554EA 181 USD
77634-554 77637-602 77654-570
Ethyl 1,3,5-trimethyl-1H-pyrazole-4-carboxylate ≥97%
Ethyl 1,3,5-trimethyl-1H-pyrazole-4-carboxylate

Precaución: For Research Use Only

Formula: C₉H₁₄N₂O₂
MW: 182.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00159644
CAS Number: 56079-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR