α,1,3-Trimethyl-1H-pyrazole-5-methanol ≥97%

Supplier: AOBChem USA

Warning

267218-5G 267218-1G 267218-500MG
77628-314EA 1484.23 USD
77628-314 77637-600 77645-576
α,1,3-Trimethyl-1H-pyrazole-5-methanol ≥97%
α,1,3-Trimethyl-1H-pyrazole-5-methanol

Precaución: For Research Use Only

Formula: 140.18 g/mol
MW: C₇H₁₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD28150308
CAS Number: 57527-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR