2-Ethoxy-3,5-dimethylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

26699-5G 26699-25G 26699-1G 26699-500MG
77632-366EA 933 USD
77632-366 77658-342 77641-630 77649-678
2-Ethoxy-3,5-dimethylbenzoic acid ≥95%
2-Ethoxy-3,5-dimethylbenzoic acid

Precaución: For Research Use Only

Formula: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD14537371
CAS Number: 1216091-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR