3-Iodo-4-nitrotoluene ≥97%

Supplier: AOBChem USA

Warning

26641-5G 26641-25G
77634-550EA 149 USD
77634-550 77654-566
3-Iodo-4-nitrotoluene ≥97%
3-Iodo-4-nitrotoluene

Precaución: For Research Use Only

Formula: C₇H₆INO₂
MW: 263.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01320681
CAS Number: 52488-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR