6-Bromo-4-methyl-2-pyridinecarboxylic acid ≥95%

Supplier: AOBChem USA

Warning

266160-5G 266160-1G 266160-500MG
77628-312EA 1484.23 USD
77628-312 77637-596 77649-674
6-Bromo-4-methyl-2-pyridinecarboxylic acid ≥95%
6-Bromo-4-methyl-2-pyridinecarboxylic acid

Precaución: For Research Use Only

Formula: 216.03 g/mol
MW: C₇H₆BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD13189137
CAS Number: 1060804-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR