1,3-Dibromo-2-fluoro-4-(trifluoromethoxy)benzene ≥95%
Supplier: AOBChem USA
|
Warning
|
266069-5G
266069-1G
266069-25G
266069-500MG
77628-310EA
1484.23
USD
77628-310
77637-594
77654-564
77649-672
1,3-Dibromo-2-fluoro-4-(trifluoromethoxy)benzene ≥95%
1,3-Dibromo-2-fluoro-4-(trifluoromethoxy)benzene
Precaución: For Research Use Only
Formula:
337.89 g/mol MW: C₇H₂Br₂F₄O Storage Temperature: Ambient |
MDL Number:
MFCD28784984 CAS Number: 1803717-02-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...