1-Bromo-2-fluoro-3-iodo-4-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

266050-5G 266050-25G 266050-1G 266050-500MG
77628-308EA 1484.23 USD
77628-308 77658-340 77641-628 77645-574
1-Bromo-2-fluoro-3-iodo-4-(trifluoromethoxy)benzene ≥97%
1-Bromo-2-fluoro-3-iodo-4-(trifluoromethoxy)benzene

Precaución: For Research Use Only

Formula: C₇H₂BrF₄IO
Storage Temperature: Ambient
MDL Number: MFCD34877602
CAS Number: 2385583-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR