3-Bromo-2-fluoro-6-(trifluoromethoxy)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

265987-5G 265987-1G 265987-500MG
77658-338EA 2399 USD
77658-338 77641-626 77645-572
3-Bromo-2-fluoro-6-(trifluoromethoxy)benzoic acid ≥95%
3-Bromo-2-fluoro-6-(trifluoromethoxy)benzoic acid

Precaución: For Research Use Only

Formula: C₈H₃BrF₄O₃
Storage Temperature: Ambient
MDL Number: MFCD34664246
CAS Number: 2090478-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR