α-Ethyl-2-methyl-4-(trifluoromethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

265946-5G 265946-1G 265946-500MG
77628-306EA 1484.23 USD
77628-306 77637-592 77649-670
α-Ethyl-2-methyl-4-(trifluoromethyl)benzenemethanol ≥95%
α-Ethyl-2-methyl-4-(trifluoromethyl)benzenemethanol

Precaución: For Research Use Only

Formula: 218.22 g/mol
MW: C₁₁H₁₃F₃O
Storage Temperature: Ambient
MDL Number: MFCD34877587
CAS Number: 1594066-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR