3-Bromo-2,5-difluoropyridine ≥97%

Supplier: AOBChem USA

Warning

26592-5G 26592-1G 26592-25G 26592-500MG
77634-548EA 282 USD
77634-548 77637-590 77654-560 77649-668
3-Bromo-2,5-difluoropyridine ≥97%
3-Bromo-2,5-difluoropyridine

Precaución: For Research Use Only

Formula: C₅H₂BrF₂N
Storage Temperature: Ambient
MDL Number: MFCD12827553
CAS Number: 1211331-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR