5-Bromo-4-methylthiophene-2-carbonitrile ≥95%

Supplier: AOBChem USA

Warning

26480-5G 26480-1G 26480-500MG 26480-25G
77634-546EA 514 USD
77634-546 77637-586 77652-552 77658-334
5-Bromo-4-methylthiophene-2-carbonitrile ≥95%
5-Bromo-4-methylthiophene-2-carbonitrile

Precaución: For Research Use Only

Formula: C₆H₄BrNS
Storage Temperature: Ambient
MDL Number: MFCD17169862
CAS Number: 304854-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR