1-(4-Fluoro-2-methoxyphenyl)-2-methyl-1-propanone ≥95%
Supplier: AOBChem USA
|
Warning
|
264746-5G
264746-25G
264746-1G
264746-500MG
77632-364EA
1180
USD
77632-364
77654-558
77641-622
77649-664
1-(4-Fluoro-2-methoxyphenyl)-2-methyl-1-propanone ≥95%
1-(4-Fluoro-2-methoxyphenyl)-2-methyl-1-propanone
Precaución: For Research Use Only
Formula:
196.22 g/mol MW: C₁₁H₁₃FO₂ Storage Temperature: Ambient |
MDL Number:
MFCD22373037 CAS Number: 1443335-86-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...