1-Propanone, 1-(5-chloro-2-methoxyphenyl)-2-methyl- ≥95%

Supplier: AOBChem USA

Warning

264741-5G 264741-25G 264741-1G 264741-500MG
77632-362EA 1196 USD
77632-362 77658-332 77641-620 77649-662
1-Propanone, 1-(5-chloro-2-methoxyphenyl)-2-methyl- ≥95%
1-Propanone, 1-(5-chloro-2-methoxyphenyl)-2-methyl-

Precaución: For Research Use Only

Formula: 212.67 g/mol
MW: C₁₁H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD14530125
CAS Number: 848997-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR