1-(2-Chloro-4-methoxyphenyl)-2-methyl-1-propanone ≥95%

Supplier: AOBChem USA

Warning

264712-5G 264712-1G 264712-500MG
77658-330EA 2064 USD
77658-330 77641-618 77645-568
1-(2-Chloro-4-methoxyphenyl)-2-methyl-1-propanone ≥95%
1-(2-Chloro-4-methoxyphenyl)-2-methyl-1-propanone

Precaución: For Research Use Only

Formula: 212.67 g/mol
MW: C₁₁H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD12139089
CAS Number: 116400-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR