1-(2,5-Dichlorophenyl)-2-methyl-1-propanone ≥95%

Supplier: AOBChem USA

Warning

264711-1G 264711-5G 264711-500MG
77637-584EA 577 USD
77637-584 77658-328 77649-660
1-(2,5-Dichlorophenyl)-2-methyl-1-propanone ≥95%
1-(2,5-Dichlorophenyl)-2-methyl-1-propanone

Precaución: For Research Use Only

Formula: 217.09 g/mol
MW: C₁₀H₁₀Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD11641805
CAS Number: 1097040-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR