1-(4-Chloro-2-methylphenyl)-2-methyl-1-propanone ≥95%

Supplier: AOBChem USA

Warning

264707-5G 264707-25G 264707-1G 264707-500MG
77632-360EA 787 USD
77632-360 77658-326 77641-616 77645-566
1-(4-Chloro-2-methylphenyl)-2-methyl-1-propanone ≥95%
1-(4-Chloro-2-methylphenyl)-2-methyl-1-propanone

Precaución: For Research Use Only

Formula: 196.67 g/mol
MW: C₁₁H₁₃ClO
Storage Temperature: Ambient
MDL Number: MFCD19314975
CAS Number: 1267004-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR